Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum tube, 50mm, outside diameter 3.15mm, inside diameter 2.39mm, wall thickness 0.38mm, 99.9% | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501850277
Encompass
Molybdenum tube, 50mm, outside diameter 3.15mm, inside diameter 2.39mm, wall thickness 0.38mm, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501850277 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF789456655EA
Molybdenum tube, 50mm, outside diameter 3.15mm, inside diameter 2.39mm, wall thickness 0.38mm, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA