Learn More
Strem, An Ascensus Company CAS# 10241-05-1. 250g. Molybdenum(V) chloride, anhydrous, 99.6%. MFCD00003467
SDP
Catalog No. 5090111710
Encompass
CAS# 10241-05-1. 250g. Molybdenum(V) chloride, anhydrous, 99.6%. MFCD00003467. Molecular Weight: 273.20. Molecular Formula: MoCl5. Color/form: dark blue to black xtl Strem# 42-1390. http://www.strem.com/catalog/v/42-1390/
Catalog No. 50-901-11710 Supplier Strem, An Ascensus Company Supplier No. 421390250G
CAS# 10241-05-1. 250g. Molybdenum(V) chloride, anhydrous, 99.6%. MFCD00003467. Molecular Weight: 273.20. Molecular Formula: MoCl5. Color/form: dark blue to black xtl Strem# 42-1390. http://www.strem.com/catalog/v/42-1390/