Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solstice Molybdenum(VI) Oxide, 99.5%, Solstice

Catalog No. 60047338 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-338 100 g Poly bottle
60-047-339 500 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-338 Supplier Solstice Supplier No. 69850100G
Add to Cart
Add to Cart

Puriss. p.a., 99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155°C
Melting Point 795°C
Color White to Yellow
CAS Max % 99.8
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.8% min
Linear Formula MoO3
UN Number NONH for all modes of transport
Solubility Information Solubility in Water - 1 g/L
Formula Weight 143.94g/mol
Percent Purity 99.5%
Manufacturing Origin USA
Odor Odorless
Packaging Poly bottle
Product Line Puriss. p.a.
Appearance Off-white to Yellowish Powder
Density 4.70g/cm3
Chemical Name or Material Molybdenum(VI) oxide, Puriss. p.a., 99.5%, Powder
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338
RTECSNumber 215-204-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.