Learn More
Strem, An Ascensus Company CAS# 1313-27-5. 10g. Molybdenum(VI) oxide (99.999%-Mo) PURATREM. MFCD00003469
SDP
Catalog No. 5090111716
Encompass
CAS# 1313-27-5. 10g. Molybdenum(VI) oxide (99.999%-Mo) PURATREM. MFCD00003469. Molecular Weight: 143.95. Molecular Formula: MoO3. Color/form: pale green pwdr. Strem# 42-1476. http://www.strem.com/catalog/v/42-1476/
Catalog No. 50-901-11716 Supplier Strem, An Ascensus Company Supplier No. 42147610G
CAS# 1313-27-5. 10g. Molybdenum(VI) oxide (99.999%-Mo) PURATREM. MFCD00003469. Molecular Weight: 143.95. Molecular Formula: MoO3. Color/form: pale green pwdr. Strem# 42-1476. http://www.strem.com/catalog/v/42-1476/