Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum(Vi) Tetrachloride Oxide

Catalog No. 3737295G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-372-95G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 37-372-95G Supplier Sigma Aldrich Supplier No. 3737295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Molybdenum(VI) tetrachloride oxide can be used:To prepare a binary metal catalyst (MoOCl4-n-BuLi) for the living polymerization of various acetylenes.As an oxidant in the aromatization of various Hantzsch-1,4-dihydropyridines.As a molybdenum (Mo) precursor to prepare nafion-Mo composite material, which is used as a catalyst for the preparation of nitrones by reacting primary amines with aldehydes.

Specifications

CAS 101385-90-4
Melting Point 258°C
Density 1.07 g/mL at 25°C
Boiling Point 115°C (0.8 mmHg, lit.)
MDL Number MFCD00066558
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.