Learn More
Electron Microscopy Sciences Molybdenum Wire, 0.20 mm, Purity 99.95% 50 FT
SDP
Catalog No. 5028057
Encompass_Preferred
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.95%
Catalog No. 50-280-57 Supplier Electron Microscopy Sciences Supplier No. 73201
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.95%