Learn More
Hach Company Molybdovanadate Reagent, 1 L
Supplier:  Hach Company 2076053
Encompass
For phosphate (phosphorus) determination by the Molybdovanadate Method: Hach method 8114. Range: up to 45.0 mg/L PO4 3-. 1 liter bottle
Catalog No. 50-199-5240