Learn More
Hach Company Molybdovanadate Reagent, 100 mL MDB
Supplier:  Hach Company 2076032
Encompass
For phosphate (phosphorus) determination by the Molybdovanadate Method: Hach method 8114. Range: up to 45.0 mg/L PO₄. 100 mL bottle
Catalog No. 50-199-5238