Learn More
Aqua Solutions MolyVer 1 Reagent Powder Pillows, 25 mL, pk/100
SDP
Catalog No. NC1586966
Encompass
For determination of high range molybdenum by the mercaptoacetic acid method (Hach method 8036). Range: up to 40.0 mg/L Mo Sample Size: 25 mL. Pack of 100 powder pillows
Catalog No. NC1586966 Supplier Aqua Solutions Supplier No. HA1414669
For determination of high range molybdenum by the mercaptoacetic acid method (Hach method 8036). Range: up to 40.0 mg/L Mo Sample Size: 25 mL. Pack of 100 powder pillows