Learn More
Hach Company MolyVer« 1 Reagent Powder Pillows, 10 mL, pk/100
Supplier:  Hach Company 2604299
Encompass
For high range molybdenum determination by the Mercaptoacetic Acid method. Sample Size: 10 mL Hach Method 8036. Pack of 100 powder pillows.
Catalog No. 50-199-6322