Learn More
Hach Company MolyVer 3 Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 1417869
Encompass
For determination of high range molybdenum by the mercaptoacetic acid method (Hach method 8036). Range: up to 40.0 mg/L Mo Sample Size: 25 mL. Pack of 100 powder pillows
Catalog No. 50-199-4946