Learn More
Medchemexpress LLC HY-10961 100mg , Momelotinib CAS:1056634-68-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10961100MG
Encompass_Preferred
HY-10961 100mg Momelotinib CAS: 1056634-68-4 Momelotinib (CYT387) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50a of 11 nM and 18 nM,respectively. CYT387 shows much less activity against JAK3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9452