Learn More
Medchemexpress LLC HY-10961 10mg Medchemexpress, Momelotinib CAS:1056634-68-4 Purity:>98%
SDP
Catalog No. 502028796
Encompass_Preferred
Medchemexpress, HY-10961 10mg Momelotinib CAS:1056634-68-4 Momelotinib (CYT387) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50a of 11 nM and 18 nM,respectively. CYT387 shows much less activity against JAK3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8796 Supplier Medchemexpress LLC Supplier No. HY1096110MG
Medchemexpress, HY-10961 10mg Momelotinib CAS:1056634-68-4 Momelotinib (CYT387) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50a of 11 nM and 18 nM,respectively. CYT387 shows much less activity against JAK3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.