Learn More
Enzo Life Sciences Momelotinib (1mg). CAS: 1056634-68-4
SDP
Catalog No. 502013517
Encompass_Preferred
Using ATP kinase assays, the IC50 values for CYT387 are a follows - JAK1 (11nM), JAK2, (18nM), JAK2 V617F (23nM) JAK3 (155nM), and TYK2 (17nM). In testing kinase arrays, CYT387 also had activity (defined as an IC50 < 100nM) detected against CDK2/cyclin A, MAPK8 (JNK1), PRKCN (PKD3), PRKD1 (PKCμ), ROCK2, and TBK1 kinase as well. Alternative name: CYT 387, N-​​(cyanomethyl)​-​​4-​​[2-​​[[4-​​(4-​​morpholinyl)​phenyl]​amino]​-​​4-​​pyrimidinyl]​-​benzamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (>20mM). Long Term Storage: -20°C.
Catalog No. 50-201-3517 Supplier Enzo Life Sciences Supplier No. ENZCHM1970001
Using ATP kinase assays, the IC50 values for CYT387 are a follows - JAK1 (11nM), JAK2, (18nM), JAK2 V617F (23nM) JAK3 (155nM), and TYK2 (17nM). In testing kinase arrays, CYT387 also had activity (defined as an IC50 < 100nM) detected against CDK2/cyclin A, MAPK8 (JNK1), PRKCN (PKD3), PRKD1 (PKCμ), ROCK2, and TBK1 kinase as well. Alternative name: CYT 387, N-​​(cyanomethyl)​-​​4-​​[2-​​[[4-​​(4-​​morpholinyl)​phenyl]​amino]​-​​4-​​pyrimidinyl]​-​benzamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (>20mM). Long Term Storage: -20°C.