Learn More
Medchemexpress LLC HY-10962 5mg Medchemexpress, Momelotinib sulfate CAS:1056636-06-6 Purity:>98%
SDP
Catalog No. 501968715
Encompass_Preferred
Medchemexpress, HY-10962 5mg Momelotinib sulfate CAS:1056636-06-6 Momelotinib sulfate (CYT387 sulfate salt) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50 of 11 nM/18 nM, 10-fold selectivity versus JAK3 (IC50=155 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8715 Supplier Medchemexpress LLC Supplier No. HY109625MG
Medchemexpress, HY-10962 5mg Momelotinib sulfate CAS:1056636-06-6 Momelotinib sulfate (CYT387 sulfate salt) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50 of 11 nM/18 nM, 10-fold selectivity versus JAK3 (IC50=155 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.