Learn More
Cayman Chemical Mometasone Furoate, Purity: ≥98%, CAS: 83919-23-7, 50 mg
SDP
Catalog No. 502980893
Encompass_Preferred
A prodrug form of glucocorticoid mometasone; reduces frequency of sneezing and nasal rubbing in a rat model of histamine-induced allergic rhinitis at 1 µg/nostril; reduces croton oil-induced ear edema in mice (ED50 = 0.31 µg/ml)
Catalog No. 50-298-0893 Supplier Cayman Chemical Supplier No. 2136550 MG
A prodrug form of glucocorticoid mometasone; reduces frequency of sneezing and nasal rubbing in a rat model of histamine-induced allergic rhinitis at 1 µg/nostril; reduces croton oil-induced ear edema in mice (ED50 = 0.31 µg/ml)