Learn More
Cayman Chemical Mometason Furoate-d3 1mg
SDP
Catalog No. 502950725
Encompass_Preferred
An internal standard for the quantification of mometasone furoate by GC- or LC-MS
Catalog No. 50-295-0725 Supplier Cayman Chemical Supplier No. 334701 MG
An internal standard for the quantification of mometasone furoate by GC- or LC-MS