Learn More
Cayman Chemical Mometason Furoate-d3 500ug
SDP
Catalog No. 502946947
Encompass_Preferred
An internal standard for the quantification of mometasone furoate by GC- or LC-MS
Catalog No. 50-294-6947 Supplier Cayman Chemical Supplier No. 33470500 UG
An internal standard for the quantification of mometasone furoate by GC- or LC-MS