Learn More
Cayman Chemical Mometasone Furoate-d3, Purity: ≥99% deuterated forms (d1-d3), 5 mg
SDP
Catalog No. 502946200
Encompass_Preferred
An internal standard for the quantification of mometasone furoate by GC- or LC-MS
Catalog No. 50-294-6200 Supplier Cayman Chemical Supplier No. 334705 MG
An internal standard for the quantification of mometasone furoate by GC- or LC-MS