Learn More
Medchemexpress LLC Mometasone furoate-13C,d6 | 83919-23-7 | 99.7% | 528.46 | 1 MG
SDP
Catalog No. 5000351716
Encompass_Preferred
Mometasone furoate-13C,d6 is a glucocorticoid receptor agonist with both anti-inflammatory and anti-allergic properties, acting as a corticosteroid agent. It is utilized for topical applications in chronic skin eczema and for managing airway inflammation in asthma. The compound functions by binding to glucocorticoid receptors, inducing conformational changes, and facilitating nuclear translocation.
  • Glucocorticoid receptor agonist.
  • Possesses anti-inflammatory and anti-allergic activity.
  • Acts as a corticosteroid agent.
  • Used for topical applications in chronic skin eczema.
  • Used in management of airway inflammation in asthma in vivo.
  • Inhibits LPS-stimulated nitrite production in J774 macrophages.
  • More potent than Dexamethasone in inhibiting iNOS expression.
  • Significantly inhibits cox-2 protein expression.
Catalog No. 50-003-51716 Supplier Medchemexpress LLC Supplier No. HY13693S21MG
Mometasone furoate-13C,d6 is a glucocorticoid receptor agonist with both anti-inflammatory and anti-allergic properties, acting as a corticosteroid agent. It is utilized for topical applications in chronic skin eczema and for managing airway inflammation in asthma. The compound functions by binding to glucocorticoid receptors, inducing conformational changes, and facilitating nuclear translocation.
  • Glucocorticoid receptor agonist.
  • Possesses anti-inflammatory and anti-allergic activity.
  • Acts as a corticosteroid agent.
  • Used for topical applications in chronic skin eczema.
  • Used in management of airway inflammation in asthma in vivo.
  • Inhibits LPS-stimulated nitrite production in J774 macrophages.
  • More potent than Dexamethasone in inhibiting iNOS expression.
  • Significantly inhibits cox-2 protein expression.