Learn More
Selleck Chemical LLC Mometasone furoate S1987-200mg
SDP
Catalog No. 503110594
Encompass_Preferred
Mometasone Furoate (SCH-32088 mometasone 17-furoate) is a glucocorticoid receptor agonist used topically to reduce inflammation of the skin or in the airways
Catalog No. 50-311-0594 Supplier Selleck Chemical LLC Supplier No. S1987200MG
Mometasone Furoate (SCH-32088 mometasone 17-furoate) is a glucocorticoid receptor agonist used topically to reduce inflammation of the skin or in the airways