Learn More
Medchemexpress LLC HY-13693 50mg Medchemexpress, Mometasone furoate CAS:83919-23-7 Purity:>98%
SDP
Catalog No. 502081467
Encompass_Preferred
Medchemexpress, HY-13693 50mg Mometasone furoate CAS:83919-23-7 Mometasone furoate (Sch32088) is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. Mometasone furoate acts as a corticosteroid agent and used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1467 Supplier Medchemexpress LLC Supplier No. HY1369350MG
Medchemexpress, HY-13693 50mg Mometasone furoate CAS:83919-23-7 Mometasone furoate (Sch32088) is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. Mometasone furoate acts as a corticosteroid agent and used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.