Learn More
Medchemexpress LLC Mometasone furoate | 83919-23-7 | 99.86% | 521.43 | 500 MG
SDP
Catalog No. 5000348808
Encompass_Preferred
Mometasone furoate (Sch32088) is a glucocorticoid receptor agonist that demonstrates anti-inflammatory and anti-allergic properties. It functions as a corticosteroid agent, employed in topical applications for the management of chronic skin eczema and airway inflammation associated with asthma in living organisms.
  • Functions as a glucocorticoid receptor agonist.
  • Possesses anti-inflammatory activity.
  • Exhibits anti-allergic activity.
  • Acts as a corticosteroid.
  • Suitable for topical applications.
  • Aids in the management of chronic skin eczema.
  • Contributes to the management of airway inflammation in asthma.
Catalog No. 50-003-48808 Supplier Medchemexpress LLC Supplier No. HY13693500MG
Mometasone furoate (Sch32088) is a glucocorticoid receptor agonist that demonstrates anti-inflammatory and anti-allergic properties. It functions as a corticosteroid agent, employed in topical applications for the management of chronic skin eczema and airway inflammation associated with asthma in living organisms.
  • Functions as a glucocorticoid receptor agonist.
  • Possesses anti-inflammatory activity.
  • Exhibits anti-allergic activity.
  • Acts as a corticosteroid.
  • Suitable for topical applications.
  • Aids in the management of chronic skin eczema.
  • Contributes to the management of airway inflammation in asthma.