Learn More
Medchemexpress LLC Mometasone furoate | 83919-23-7 | 99.9% | 521.43 | 100 MG
SDP
Catalog No. 5000351736
Encompass_Preferred
Mometasone furoate is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. It is used in topical applications for chronic skin eczema and for managing airway inflammation in asthma.
  • Glucocorticoid receptor agonist
  • Anti-inflammatory and anti-allergic activity
  • Corticosteroid agent
  • Used for topical applications in chronic skin eczema
  • Used for airway inflammation management of asthma in vivo
Catalog No. 50-003-51736 Supplier Medchemexpress LLC Supplier No. HY13693100MG
Mometasone furoate is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. It is used in topical applications for chronic skin eczema and for managing airway inflammation in asthma.
  • Glucocorticoid receptor agonist
  • Anti-inflammatory and anti-allergic activity
  • Corticosteroid agent
  • Used for topical applications in chronic skin eczema
  • Used for airway inflammation management of asthma in vivo