Learn More
Medchemexpress LLC Mometasone furoate | 83919-23-7 | MFCD00866003 | 99.86% | 521.43 | 200 MG
SDP
Catalog No. 5000350710
Encompass_Preferred
Mometasone furoate is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. It acts as a corticosteroid agent used for topical applications in chronic skin eczema and airway inflammation management of asthma. It binds to a glucocorticoid receptor, causing conformational changes and separation from chaperones, and the receptor moves to the nucleus.
  • Strongly inhibits LPS-stimulated nitrite production in J774 macrophages
  • More potent than DEX in inhibiting iNOS expression
  • Significant inhibition of cox-2 protein expression
Catalog No. 50-003-50710 Supplier Medchemexpress LLC Supplier No. HY13693200MG
Mometasone furoate is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity. It acts as a corticosteroid agent used for topical applications in chronic skin eczema and airway inflammation management of asthma. It binds to a glucocorticoid receptor, causing conformational changes and separation from chaperones, and the receptor moves to the nucleus.
  • Strongly inhibits LPS-stimulated nitrite production in J774 macrophages
  • More potent than DEX in inhibiting iNOS expression
  • Significant inhibition of cox-2 protein expression