Learn More
Medchemexpress LLC Mometasone furoate | 83919-23-7 | MFCD00866003 | 99.91% | 521.43 | 25 MG
SDP
Catalog No. 5000350631
Encompass_Preferred
Mometasone furoate (Standard) is an analytical standard primarily used for research and analytical applications. This glucocorticoid receptor agonist exhibits anti-inflammatory and anti-allergic activity, functioning as a corticosteroid agent for topical applications. It is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Glucocorticoid receptor agonist
  • Exhibits anti-inflammatory and anti-allergic activity
  • Functions as a corticosteroid agent
  • Used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
Catalog No. 50-003-50631 Supplier Medchemexpress LLC Supplier No. 25MGHY13693R
Mometasone furoate (Standard) is an analytical standard primarily used for research and analytical applications. This glucocorticoid receptor agonist exhibits anti-inflammatory and anti-allergic activity, functioning as a corticosteroid agent for topical applications. It is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Glucocorticoid receptor agonist
  • Exhibits anti-inflammatory and anti-allergic activity
  • Functions as a corticosteroid agent
  • Used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS