Learn More
Medchemexpress LLC Mometasone furoate | MFCD00866003 | 99.9% | 521.43 g/mol | C27H30Cl2O6 | 5 MG
SDP
Catalog No. 5000221914
Encompass_Preferred
Mometasone furoate-13C,d6 is an isotopically labeled reference standard of mometasone furoate intended for analytical and research use. It provides a stable, labeled analog for method development and quantitative mass spectrometry studies involving glucocorticoid receptor assays. Parent compound properties: CAS 83919-23-7; formula C27H30Cl2O6; molecular weight 521.43 g/mol.
  • Isotopically labeled with 13C and six deuteriums for clear mass separation.
  • Suitable as an internal standard for quantitative LC-MS/MS assays.
  • Chemically analogous to the parent compound for accurate recovery and matrix matching.
  • Intended for analytical, reference, and method development applications.
Catalog No. 50-002-21914 Supplier Medchemexpress LLC Supplier No. HY13693S25MG
Mometasone furoate-13C,d6 is an isotopically labeled reference standard of mometasone furoate intended for analytical and research use. It provides a stable, labeled analog for method development and quantitative mass spectrometry studies involving glucocorticoid receptor assays. Parent compound properties: CAS 83919-23-7; formula C27H30Cl2O6; molecular weight 521.43 g/mol.
  • Isotopically labeled with 13C and six deuteriums for clear mass separation.
  • Suitable as an internal standard for quantitative LC-MS/MS assays.
  • Chemically analogous to the parent compound for accurate recovery and matrix matching.
  • Intended for analytical, reference, and method development applications.