Learn More
Medchemexpress LLC Mometasone furoate (Standard) | 83919-23-7 | 99.9% | 521.43 | 50 MG
SDP
Catalog No. 5000352054
Encompass_Preferred
Mometasone furoate (Standard) is the analytical standard of Mometasone furoate, intended for research and analytical applications. It is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity.
  • Acts as a corticosteroid agent
  • Used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo
  • Grade of analytical standard, which is the reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-003-52054 Supplier Medchemexpress LLC Supplier No. HY13693R50MG
Mometasone furoate (Standard) is the analytical standard of Mometasone furoate, intended for research and analytical applications. It is a glucocorticoid receptor agonist with anti-inflammatory and anti-allergic activity.
  • Acts as a corticosteroid agent
  • Used for topical applications in chronic skin eczema and airway inflammation management of asthma in vivo
  • Grade of analytical standard, which is the reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS