Learn More
Selleck Chemical LLC MOMIPP E2933-25MG
SDP
Catalog No. 5000040121
Encompass_Preferred
MOMIPP a macropinocytosis inducer is a PIKfyve inhibitor MOMIPP readily penetrates the blood-brain barrier and is moderately effective in suppressing progression of intracerebral glioblastoma xenografts
Catalog No. 50-000-40121 Supplier Selleck Chemical LLC Supplier No. E293325MG
MOMIPP a macropinocytosis inducer is a PIKfyve inhibitor MOMIPP readily penetrates the blood-brain barrier and is moderately effective in suppressing progression of intracerebral glioblastoma xenografts