Learn More
Medchemexpress LLC Momordicin IV | 894412-35-2 | 634.84 | 1 MG
SDP
Catalog No. 5000416041
Encompass_Preferred
Momordicin IV is a natural product classified under Terpenoids and Triterpenes. It is initially sourced from plants, specifically from the Cucurbitaceae family, Momordica charantia Linn. This product is for research use only.
  • 98% purity
  • Solid appearance
  • White to off-white color
  • Ships at room temperature (continental US)
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)
Catalog No. 50-004-16041 Supplier Medchemexpress LLC Supplier No. HYN135791MG
Momordicin IV is a natural product classified under Terpenoids and Triterpenes. It is initially sourced from plants, specifically from the Cucurbitaceae family, Momordica charantia Linn. This product is for research use only.
  • 98% purity
  • Solid appearance
  • White to off-white color
  • Ships at room temperature (continental US)
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)