Learn More
New England Biolabs, Inc. Monarch® gDNA Binding Buffer - 65 ml
SDP
Catalog No. 501959400
Encompass
The Monarch gDNA Binding Buffer is a chaotropic salt-based binding buffer with low alcohol content, designed to optimize binding of gDNA to the spin column. The binding of gDNA takes place at low speed, maximizing binding and efficiency. This buffer is a component of the Monarch Genomic DNA Purification Kit.
  • A component of the Monarch Genomic DNA Purification Kit
  • A chaotropic salt-based binding buffer with low alcohol content for optimal binding of gDNA
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-195-9400 Supplier New England Biolabs, Inc. Supplier No. T3014L
The Monarch gDNA Binding Buffer is a chaotropic salt-based binding buffer with low alcohol content, designed to optimize binding of gDNA to the spin column. The binding of gDNA takes place at low speed, maximizing binding and efficiency. This buffer is a component of the Monarch Genomic DNA Purification Kit.
  • A component of the Monarch Genomic DNA Purification Kit
  • A chaotropic salt-based binding buffer with low alcohol content for optimal binding of gDNA
  • Monarch kit components are also available separately for added convenience