Learn More
New England Biolabs, Inc. Monarch® gDNA Blood Lysis Buffer - 20 ml
SDP
Catalog No. 501959399
Encompass
The Monarch gDNA Blood Lysis Buffer contains a strong protective component against the high nuclease activity in blood samples and supports rapid degradation of hemoglobin and other protein components. This buffer is a component of the Monarch Genomic DNA Purification Kit.
  • A component of the Monarch Genomic DNA Purification Kit
  • Supports rapid degradation of hemoglobin and other protein components
  • Contains a strong protective component against the high nuclease activity in blood samples
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-195-9399 Supplier New England Biolabs, Inc. Supplier No. T3013L
The Monarch gDNA Blood Lysis Buffer contains a strong protective component against the high nuclease activity in blood samples and supports rapid degradation of hemoglobin and other protein components. This buffer is a component of the Monarch Genomic DNA Purification Kit.
  • A component of the Monarch Genomic DNA Purification Kit
  • Supports rapid degradation of hemoglobin and other protein components
  • Contains a strong protective component against the high nuclease activity in blood samples
  • Monarch kit components are also available separately for added convenience