Learn More
New England Biolabs, Inc. Monarch® gDNA Cell Lysis Buffer - 20 ml
SDP
Catalog No. 501959398
Encompass
The Monarch gDNA Cell Lysis Buffer is a component of the Monarch Genomic DNA Purification Kit . It provides mild lysis conditions that help to reduce the viscosity common in cell samples. Optimized for speed and efficiency, this buffer requires only a 5 minute lysis incubation time with Monarch Proteinase K and Monarch RNase A to obtain maximal yields of highly pure and intact gDNA with a typical peak size >60 kb.
  • A component of the Monarch Genomic DNA Purification Kit
  • Provides mild lysis conditions that help to reduce the viscosity common in cell samples
  • Requires only a 5 minute lysis incubation with Monarch Proteinase K and Monarch RNase A
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-195-9398 Supplier New England Biolabs, Inc. Supplier No. T3012L
The Monarch gDNA Cell Lysis Buffer is a component of the Monarch Genomic DNA Purification Kit . It provides mild lysis conditions that help to reduce the viscosity common in cell samples. Optimized for speed and efficiency, this buffer requires only a 5 minute lysis incubation time with Monarch Proteinase K and Monarch RNase A to obtain maximal yields of highly pure and intact gDNA with a typical peak size >60 kb.
  • A component of the Monarch Genomic DNA Purification Kit
  • Provides mild lysis conditions that help to reduce the viscosity common in cell samples
  • Requires only a 5 minute lysis incubation with Monarch Proteinase K and Monarch RNase A
  • Monarch kit components are also available separately for added convenience