Learn More
New England Biolabs, Inc. Monarch® gDNA Elution Buffer - 34 ml
SDP
Catalog No. 501959402
Encompass
Monarch gDNA Elution Buffer is supplied in 10 mM Tris-HCl, pH 9.0, 0.1 mM EDTA and is suitable for long term storage of gDNA. Nucleases are inactivated both by the inclusion of EDTA and the pH of the solution. This buffer is a component of the Monarch Genomic DNA Purification Kit.
  • A component of the Monarch Genomic DNA Purification Kit
  • 10 mM Tris-HCl, pH 9.0, 0.1 mM EDTA
  • Suitable for long term storage of gDNA
  • Nucleases are inactivated both by the inclusion of EDTA and the pH of the solution
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-195-9402 Supplier New England Biolabs, Inc. Supplier No. T3016L
Monarch gDNA Elution Buffer is supplied in 10 mM Tris-HCl, pH 9.0, 0.1 mM EDTA and is suitable for long term storage of gDNA. Nucleases are inactivated both by the inclusion of EDTA and the pH of the solution. This buffer is a component of the Monarch Genomic DNA Purification Kit.
  • A component of the Monarch Genomic DNA Purification Kit
  • 10 mM Tris-HCl, pH 9.0, 0.1 mM EDTA
  • Suitable for long term storage of gDNA
  • Nucleases are inactivated both by the inclusion of EDTA and the pH of the solution
  • Monarch kit components are also available separately for added convenience