Learn More
New England Biolabs, Inc. Monarch® gDNA Purification Columns - 100 columns
SDP
Catalog No. 501959403
Encompass
The Monarch gDNA Purification Columns are a component of the Monarch Genomic DNA Purification Kit and can be used to purify up to 30 ug genomic DNA from a variety of sample types. They are available for purchase separately for your convenience and supplied with Monarch Collection Tubes II.
  • A component of the Monarch Genomic DNA Purification Kit
  • Can be used to purify up to 30 ug genomic DNA from a variety of sample types
  • Contains 100 columns and 100 collection tubes
  • Compatible with Monarch Collection Tubes II
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-195-9403 Supplier New England Biolabs, Inc. Supplier No. T3017L
The Monarch gDNA Purification Columns are a component of the Monarch Genomic DNA Purification Kit and can be used to purify up to 30 ug genomic DNA from a variety of sample types. They are available for purchase separately for your convenience and supplied with Monarch Collection Tubes II.
  • A component of the Monarch Genomic DNA Purification Kit
  • Can be used to purify up to 30 ug genomic DNA from a variety of sample types
  • Contains 100 columns and 100 collection tubes
  • Compatible with Monarch Collection Tubes II
  • Monarch kit components are also available separately for added convenience