Learn More
New England Biolabs, Inc. Monarch® gDNA Tissue Lysis Buffer - 34 ml
SDP
Catalog No. 501959397
Encompass
The Monarch Tissue Lysis Buffer is a component of the Monarch Genomic DNA Purification Kit. It provides lysis conditions that enable rapid digestion of tissue pieces while simultaneously ensuring the genomic DNA fragment length is optimal for binding and elution. Coupled with optimized Proteinase K digestion, the tissue lysis system facilitates above-average yields for common animal tissues , including tissues that prove difficult for many other commercial kits (e.g., brain and muscle).
  • A component of the Monarch Genomic DNA Purification Kit
  • Provides lysis conditions for rapid digestion of tissue while ensuring optimal DNA fragment length
  • Coupled with Proteinase K digestion, enables above-average yields for common animal tissues
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-195-9397 Supplier New England Biolabs, Inc. Supplier No. T3011L
The Monarch Tissue Lysis Buffer is a component of the Monarch Genomic DNA Purification Kit. It provides lysis conditions that enable rapid digestion of tissue pieces while simultaneously ensuring the genomic DNA fragment length is optimal for binding and elution. Coupled with optimized Proteinase K digestion, the tissue lysis system facilitates above-average yields for common animal tissues , including tissues that prove difficult for many other commercial kits (e.g., brain and muscle).
  • A component of the Monarch Genomic DNA Purification Kit
  • Provides lysis conditions for rapid digestion of tissue while ensuring optimal DNA fragment length
  • Coupled with Proteinase K digestion, enables above-average yields for common animal tissues
  • Monarch kit components are also available separately for added convenience