Learn More
New England Biolabs, Inc. Monarch® gDNA Wash Buffer - 60 ml
SDP
Catalog No. 501959401
Encompass
Monarch gDNA Wash Buffer is a component of the Monarch Genomic DNA Purification Kit. It is an ethanol-based, low salt buffer that allows for complete removal of chaotropic salt and cell components in just 2 wash steps, enabling the purification of highly-pure gDNA.
  • A component of the Monarch Genomic DNA Purification Kit
  • An ethanol-based, low salt buffer that allows for removal of chaotropic salt and cell components
  • Supplied as a concentrate
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-195-9401 Supplier New England Biolabs, Inc. Supplier No. T3015L
Monarch gDNA Wash Buffer is a component of the Monarch Genomic DNA Purification Kit. It is an ethanol-based, low salt buffer that allows for complete removal of chaotropic salt and cell components in just 2 wash steps, enabling the purification of highly-pure gDNA.
  • A component of the Monarch Genomic DNA Purification Kit
  • An ethanol-based, low salt buffer that allows for removal of chaotropic salt and cell components
  • Supplied as a concentrate
  • Monarch kit components are also available separately for added convenience