Learn More
Aviva Systems Biology Corp MON1A Antibody - C-terminal region (ARP60472_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60472_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MON1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MON1A. Alternate names: FLJ97088, MGC13272, SAND1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-052