Learn More
Aviva Systems Biology Corp MON2 Antibody - C-terminal region (ARP65788_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65788_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MON2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human MON2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rat
Catalog No. 50-288-573