Learn More
Cayman Chemical MonacylglycRol LIpase InhIb 96
SDP
Catalog No. 502893538
Encompass_Preferred
MAGL is the main enzyme responsible for the inactivation of 2-AG. Cayman’s Monoacylglycerol Lipase Inhibitor Screening Assay provides a convenient method for screening human MAGL inhibitors. MAGL hydrolyzes 4-nitrophenylacetate resulting in a yellow product, 4-nitrophenol, with an absorbance of 405-412 nm.
Catalog No. 50-289-3538 Supplier Cayman Chemical Supplier No. 70519296 WELL
MAGL is the main enzyme responsible for the inactivation of 2-AG. Cayman’s Monoacylglycerol Lipase Inhibitor Screening Assay provides a convenient method for screening human MAGL inhibitors. MAGL hydrolyzes 4-nitrophenylacetate resulting in a yellow product, 4-nitrophenol, with an absorbance of 405-412 nm.