Learn More
Cayman Chemical Monoacylglycerol Lipase Inhibitor 21, Purity: ≥98%, CAS: 1643657-35-5, 10 mg
SDP
Catalog No. 502967795
Encompass_Preferred
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively); does not inhibit ABHD6, ABHD12, CB1, or CB2 receptors (Kis > 10 µM); used to ameliorate disease progression in a mouse model of multiple sclerosis
Catalog No. 50-296-7795 Supplier Cayman Chemical Supplier No. 1696410 MG
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively); does not inhibit ABHD6, ABHD12, CB1, or CB2 receptors (Kis > 10 µM); used to ameliorate disease progression in a mouse model of multiple sclerosis