Learn More
Cayman Chemical MonacylglycRol lpase InHB 50mg
SDP
Catalog No. 502968277
Encompass_Preferred
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively); does not inhibit ABHD6, ABHD12, CB1, or CB2 receptors (Kis > 10 µM); used to ameliorate disease progression in a mouse model of multiple sclerosis
Catalog No. 50-296-8277 Supplier Cayman Chemical Supplier No. 1696450 MG
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively); does not inhibit ABHD6, ABHD12, CB1, or CB2 receptors (Kis > 10 µM); used to ameliorate disease progression in a mouse model of multiple sclerosis