Learn More
New England Biolabs, Inc. Monarch® Buffer BX - 80 ml
SDP
Catalog No. NC1738741
Encompass
The Monarch Buffer BX, a component of Monarch Nucleic Acid Purification Kits, is a guanidine-based buffer designed for binding steps in nucleic acid purification.
  • A component of the Monarch RNA Cleanup Kits
  • A guanidine-based buffer designed for binding steps in nucleic acid purification
  • Monarch kit components are also available separately for added convenience
  • Monarch kit components are also available separately for added convenience
Catalog No. NC1738741 Supplier New England Biolabs, Inc. Supplier No. T2041L
The Monarch Buffer BX, a component of Monarch Nucleic Acid Purification Kits, is a guanidine-based buffer designed for binding steps in nucleic acid purification.
  • A component of the Monarch RNA Cleanup Kits
  • A guanidine-based buffer designed for binding steps in nucleic acid purification
  • Monarch kit components are also available separately for added convenience
  • Monarch kit components are also available separately for added convenience