Learn More
New England Biolabs, Inc. Monarch® 2 ml Tubes - 100 tubes
SDP
Catalog No. 502068306
Encompass
The Monarch 2 ml Tubes are supplied as a component of the Monarch HMW DNA Extraction Kits. The tubes are round-bottom and have a locking mechanism for safe and convenient handling. The tubes are low DNA binding, making them optimal for purification of HMW DNA in the Monarch workflows.
  • A component of the Monarch HMW DNA Extraction Kits
  • Low DNA binding
  • Includes locking mechanism on the cap
  • 2 ml capacity
Catalog No. 50-206-8306 Supplier New England Biolabs, Inc. Supplier No. T3003L
The Monarch 2 ml Tubes are supplied as a component of the Monarch HMW DNA Extraction Kits. The tubes are round-bottom and have a locking mechanism for safe and convenient handling. The tubes are low DNA binding, making them optimal for purification of HMW DNA in the Monarch workflows.
  • A component of the Monarch HMW DNA Extraction Kits
  • Low DNA binding
  • Includes locking mechanism on the cap
  • 2 ml capacity