Learn More
New England Biolabs, Inc. Monarch® Bead Retainers - 100 pieces
SDP
Catalog No. 502068308
Encompass
Plastic parts for retaining the Monarch DNA Capture Beads during the Monarch HMW DNA extraction workflow. These pieces resemble spin columns without any membrane inserted and facilitate various steps of the extraction workflow. They mate to the Monarch Collection Tube II.
  • Supplied as a component of the Monarch HMW DNA Extraction Kits
  • Facilitates Monarch HMW DNA extraction workflow by containing the beads during wash and elution
  • Plastic part resembling a spin column without a membrane
Catalog No. 50-206-8308 Supplier New England Biolabs, Inc. Supplier No. T3004L
Plastic parts for retaining the Monarch DNA Capture Beads during the Monarch HMW DNA extraction workflow. These pieces resemble spin columns without any membrane inserted and facilitate various steps of the extraction workflow. They mate to the Monarch Collection Tube II.
  • Supplied as a component of the Monarch HMW DNA Extraction Kits
  • Facilitates Monarch HMW DNA extraction workflow by containing the beads during wash and elution
  • Plastic part resembling a spin column without a membrane