Learn More
New England Biolabs, Inc. Monarch® Buffer B1 - 57 ml
SDP
Catalog No. 503025702
Encompass
Monarch Buffer B1 is included in Monarch Nucleic Acid Purification Kits, and also offered as a standalone buffer for convenience. It is supplied without RNase A. Refer to the appropriate buffer prepration instructions for your specific workflow before use.
  • A component of the Monarch DNA & RNA Purification Kits, also offered individually for convenience
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
  • Supplied without RNase A
  • Monarch Buffers and Kits are designed with significantly less plastic than leading suppliers
Catalog No. 50-302-5702 Supplier New England Biolabs, Inc. Supplier No. T1111L
Monarch Buffer B1 is included in Monarch Nucleic Acid Purification Kits, and also offered as a standalone buffer for convenience. It is supplied without RNase A. Refer to the appropriate buffer prepration instructions for your specific workflow before use.
  • A component of the Monarch DNA & RNA Purification Kits, also offered individually for convenience
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
  • Supplied without RNase A
  • Monarch Buffers and Kits are designed with significantly less plastic than leading suppliers