Learn More
New England Biolabs, Inc. Monarch® Buffer BY - 230 ml
SDP
Catalog No. 503025700
Encompass
Monarch Buffer BY is included in Monarch Nucleic Acid Purification Kits, and also offered as a standalone buffer for convenience. This buffer is designed for use with multiple kits and workflows to provide maximum value and flexibility. Refer to the appropriate guidance of the corresponding kit for instructions on preparing the buffer for use in a specific workflow. This buffer is used to dissolve the agarose containing the target DNA and create an optimal condition for DNA to effectively bind to the spin columns.
  • A component of the Monarch DNA & RNA Purification Kits, also offered individually for convenience
  • This buffer may be used in multiple kits and workflows
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
Catalog No. 50-302-5700 Supplier New England Biolabs, Inc. Supplier No. T1121L
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Monarch Buffer BY is included in Monarch Nucleic Acid Purification Kits, and also offered as a standalone buffer for convenience. This buffer is designed for use with multiple kits and workflows to provide maximum value and flexibility. Refer to the appropriate guidance of the corresponding kit for instructions on preparing the buffer for use in a specific workflow. This buffer is used to dissolve the agarose containing the target DNA and create an optimal condition for DNA to effectively bind to the spin columns.
  • A component of the Monarch DNA & RNA Purification Kits, also offered individually for convenience
  • This buffer may be used in multiple kits and workflows
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow