Learn More
New England Biolabs, Inc. Monarch® Buffer WZ - 26 ml
SDP
Catalog No. 503025706
Encompass
Monarch Buffer WZ is included in Monarch Nucleic Acid Purification Kits, and also offered as a standalone buffer for convenience. This buffer is designed for use with multiple kits and workflows to provide maximum value and flexibility. Refer to the appropriate guidance of the corresponding kit for instructions on preparing the buffer for use in a specific workflow. This is designed as a wash buffer to remove any trace buffer components and salts.
  • A component of the Monarch DNA & RNA Purification Kits, also offered individually for convenience
  • This buffer may be used in multiple kits and workflows
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
  • Supplied as a concentrate, add appropriate alcohol before use
Catalog No. 50-302-5706 Supplier New England Biolabs, Inc. Supplier No. T1115L
Monarch Buffer WZ is included in Monarch Nucleic Acid Purification Kits, and also offered as a standalone buffer for convenience. This buffer is designed for use with multiple kits and workflows to provide maximum value and flexibility. Refer to the appropriate guidance of the corresponding kit for instructions on preparing the buffer for use in a specific workflow. This is designed as a wash buffer to remove any trace buffer components and salts.
  • A component of the Monarch DNA & RNA Purification Kits, also offered individually for convenience
  • This buffer may be used in multiple kits and workflows
  • Refer to appropriate instructions on preparing the buffer for use in a specific workflow
  • Supplied as a concentrate, add appropriate alcohol before use