Learn More
New England Biolabs, Inc. Monarch® DNA Capture Beads - 200 beads
SDP
Catalog No. 502068307
Encompass
These specialized glass beads are optimized for capturing HMW DNA in the Monarch HMW DNA workflows.
  • A component of the Monarch HMW DNA Extraction Kits
  • 4 mm borosilicate glass beads
  • Optimized for extraction of HMW DNA
  • Smooth surface to maximize elution efficiency
  • Easy to handle when coupled with the Monarch Bead Retainers
Catalog No. 50-206-8307 Supplier New England Biolabs, Inc. Supplier No. T3005L
These specialized glass beads are optimized for capturing HMW DNA in the Monarch HMW DNA workflows.
  • A component of the Monarch HMW DNA Extraction Kits
  • 4 mm borosilicate glass beads
  • Optimized for extraction of HMW DNA
  • Smooth surface to maximize elution efficiency
  • Easy to handle when coupled with the Monarch Bead Retainers