Learn More
New England Biolabs, Inc. Monarch DNA Gel Extraction Kit – 50 preps
SDP
Supplier:  New England Biolabs, Inc. T1020S
Encompass
The Monarch DNA Gel Extraction Kit rapidly and reliably purifies up to 5 ug of concentrated, high-quality DNA from agarose gels. The kit utilizes a bind/wash/elute workflow with minimal incubation and spin times. The columns ensure zero buffer retention and no carryover of contaminants, enabling elution of sample in volumes as low as 6 ul. The buffers provided have been optimized, and do not require monitoring of pH. Unlike other kits, there is no need to add isopropanol to the melted agarose prior to loading on the column, saving you a step. Eluted DNA is ready for use in restriction digests, DNA sequencing, ligation and other enzymatic manipulations. Designed with sustainability in mind, Monarch kits use significantly less plastic and responsibly-sourced, recyclable packaging.
Catalog No. NC1197380